Learn More
Curcumin, 95% (total curcuminoid content), from Turmeric rhizome, Thermo Scientific Chemicals
£40.30 - £491.45
Chemical Identifiers
CAS | 458-37-7 |
---|---|
Molecular Formula | C21H20O6 |
Molecular Weight (g/mol) | 368.39 |
MDL Number | MFCD00008365 |
InChI Key | ZIUSSTSXXLLKKK-KOBPDPAPSA-N |
Synonym | curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i |
PubChem CID | 969516 |
ChEBI | CHEBI:3962 |
IUPAC Name | (1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione |
SMILES | COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11417174
|
Thermo Scientific Alfa Aesar
B21573.09 |
10 g |
£40.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427174
|
Thermo Scientific Alfa Aesar
B21573.18 |
50 g |
£124.62
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437174
|
Thermo Scientific Alfa Aesar
B21573.30 |
250 g |
£491.45
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Curcumin is used for boron quantification. It inhibits lipoxygenase and cyclooxygenase. It acts as a dietary supplement and is used in pharmaceutical industry. It acts as a food coloring agent and acts as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCurcumin is used for boron quantification. It inhibits lipoxygenase and cyclooxygenase. It acts as a dietary supplement and is used in pharmaceutical industry. It acts as a food coloring agent and acts as a food additive.
Solubility
Soluble in acetone, dimethylsufoxide, ethanol and dimethyl formamide. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
458-37-7 | |
368.39 | |
ZIUSSTSXXLLKKK-KOBPDPAPSA-N | |
969516 | |
(1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione |
C21H20O6 | |
MFCD00008365 | |
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i | |
CHEBI:3962 | |
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O |
Specifications
458-37-7 | |
0.95 | |
MFCD00008365 | |
14,2673 | |
Soluble in acetone,dimethylsufoxide,ethanol and dimethyl formamide. Insoluble in water. | |
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O | |
368.39 | |
CHEBI:3962 | |
95% | |
Curcumin, from Turmeric rhizome |
170°C to 175°C | |
C21H20O6 | |
2306965 | |
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i | |
ZIUSSTSXXLLKKK-KOBPDPAPSA-N | |
(1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione | |
969516 | |
368.39 | |
(total curcuminoid content) |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 207-280-5
missing translation for 'rtecsNumber' : MI5230000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Curcumin, 95% (total curcuminoid content), from Turmeric rhizome
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 1,6-Heptadiene-3,5-dione, 1,7-bis(4-hydroxy-3-methoxyphenyl)-, (E,E)-
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.