missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals Product Code.: 10368110

Thermo Scientific Chemicals Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals

Product Code. 10368110
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10368110

Brand: Thermo Scientific Chemicals 218580500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458-37-7
Molecular Formula C21H20O6
Molecular Weight (g/mol) 368.39
MDL Number MFCD00008365
InChI Key ZIUSSTSXXLLKKK-KOBPDPAPSA-N
Synonym curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
PubChem CID 969516
ChEBI CHEBI:3962
SMILES COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O

Specifications

Melting Point 183.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula [HOC6H3(OCH3)CH=CHCO]2CH2
Refractive Index 1.4155 to 1.4175
Quantity 50 g
Beilstein 08,IV,3697
Merck Index 15,2663
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in diethyl ether,soluble in alcohol,glacial acetic acid,acetone
IUPAC Name (1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
Formula Weight 368.38
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462571 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number218580500,218580100,218582500
Lot NumberA0457737 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number218580500,218580100,218582500
Lot NumberA0453300 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number218580500,218580100,218582500
Lot NumberA0454969 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number218580500,218580100,218582500
Lot NumberA0441759 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number218580500,218580100,218582500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.