missing translation for 'onlineSavingsMsg'
Learn More

Curdione, MedChemExpress

MedChemExpress Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.

Encompass_Preferred

£102.85 - £287.98

Specifications

Chemical Name or Material Curdione
CAS 13657-68-6
Color White
Purity Grade Notes Research
Molecular Formula C15H24O2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16474817
Encompass_Preferred
View Documents
Medchem Express
HY-N0353
50 mg
£287.98
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16408958
Encompass_Preferred
View Documents
Medchem Express
HY-N0353
10 mg
£102.85
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.
Specifications

Specifications

Curdione
White
C15H24O2
DMSO : ≥ 100 mg/mL (423.10 mM)
236.35
98.61%
Solid
13657-68-6
Research
(+)-Curdione
O=C1C[C@@H](C(C)C)C(C/C(C)=C/CC[C@@H]1C)=O
236.35
Research
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Curdione, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.