missing translation for 'onlineSavingsMsg'
Learn More

Notrax Cushion Flex™ Oil-Resistant Diamond Plate Anti-Fatigue Mat

Notrax Cushion Flex™ - Oil-resistant version of our bestselling anti-fatigue mat best known for its diamond plate top surface and high level of ergonomic comfort in both dynamic and stationary working positions.

Brand:  Notrax 489S002MBY

 View more versions of this product

Product Code. 17762178

  • £399.00 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The oil-resistant version of our bestselling anti-fatigue mat best known for its diamond plate top surface and high level of ergonomic comfort in both dynamic and stationary working positions
  • A 12
  • 7 mm thick worker platform offers maximum durability, comfort, and insulation from hard floors and vibrations
  • 100% nitrile rubber compound is resistant to most industrial oils
  • Slip resistance R10 according to DIN51130 and BG-rule BGR181
  • Yellow bevelled edges on all four sides minimize risk of tripping and comply with OSHA code 1910-144
Specifications

Specifications

Black/Yellow
210 cm
Industrial and Commercial Matting
6.9 ft.
Nitrile Rubber
91 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.