missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Pairs
Custom 5' Fluorescent Labeled/Unlabeled Di-repeat + Tail
£144.00 - £350.00
Specifications
Product Type | Custom 5' Labeled or Unlabeled Di-Repeat + Tail |
---|---|
Content And Storage | -20°C |
For Use With (Application) | Fragment Analysis, Gene Expression Studies, Genotyping, PCR |
Form | Delivered either in Solution or Dried down |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15557240
|
Applied Biosystems™
4304979 |
10,000 pmol |
£144.00
10000pmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15597240
|
Applied Biosystems™
4304981 |
80,000 pmol |
£230.00
80000pmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15577280
|
Applied Biosystems™
4304982 |
300,000 pmol |
£350.00
300000pmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.Specifications
Custom 5' Labeled or Unlabeled Di-Repeat + Tail | |
-20°C | |
Fragment Analysis, Gene Expression Studies, Genotyping, PCR | |
Delivered either in Solution or Dried down |
Safety and Handling
missing translation for 'shelfLife' : 12 Months from Manufacturing Date
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title