Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Primer Pairs
Custom 5' Fluorescent Labeled/Unlabeled Pairs
Brand:  Applied Biosystems™ 450056
Product Code. 15419192
  • £147.00 / Each
Explore more special offers
Description
Description
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Custom 5' Fluorescent Labeled/Unlabeled Pairs, 10000pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, 12 Months Shelf Life, –20°C Storage Temperature, Desalted Purification, Solution or Dried Down Format, For Use in Fragment Analysis, Gene Expression Studies, Genotyping and PCR
TE or Water
5' Labeled
Fragment Analysis, Gene Expression Studies, Genotyping, PCR
Custom 5' Fluorescent Labeled or Unlabeled Pairs
-20°C
10,000 pmol
Solution or Dried down