missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Custom Cyclic Peptide Microarray

Custom peptide microarray with up to 5500 different cyclic constrained peptides printed in duplicate. The microarray covers any custom epitope, peptide or antigen collection for antibody validation, epitope mapping, serum screening, biomarker discovery or protein-peptide interaction studies.

Brand:  PEPperCHIP™ PPC.014.001

Additional Details : Weight : 0.02200kg

Product Code. 16307748

  • £1319.70 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: Custom antigen cyclic constrained peptides, epitope or peptide collections
  • Microarray Layout: 1, 2, 3, 4 , 5 or 16 array copies per microarray
  • Available Amino Acids: L- and D-amino acids, citrulline, acetyllysine and other amino acids with posttranslational modifications
  • Peptide Lengths: Variable with routine synthesis of 7, 10 and 13mer peptides, longer peptides on request
  • Control Peptides: HA, polio and/or flag control peptides, custom controls on request
  • Microarray Format: 75.4 x 25 mm
Specifications

Specifications

PEPperSlide™ PEGMA/ PMMA, Cystein Pre-attachment
Antibody validation, epitope mapping, serum screening, biomarker discovery, protein-peptide interaction studies
Microscopic Glass
Individual barcode for each microarray
Combinatorial peptide synthesis by amino acid laser printing and thioether cyclization
ISO 9001:2015
RUO
75.4 x 25 mm
Peptide Microarray
Non-sterile
1 Slide
1 mm
Transparent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers