missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Custom Designed Ambion™ In Vivo
Click to view available options
:
5 nmol
Description
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Specifications
| Color | White |
| Content And Storage | Room temperature |
| Description | Custom Ambion™ In-vivo Pre-designed siRNA, 1umol, Solid, Powder, White, Standard Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA |
| For Use With (Application) | RNAi, epigenetics and non-coding RNA research, RNAi, siRNA |
| Product Type | Custom Ambion™ In-vivo Pre-designed siRNA |
| Purity | Standard |
| Quantity | 1 umol |
| Form | Solid |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction