Learn More
Ambion™ Custom mirVana™ miRNA Inhibitor
Custom mirVana™ miRNA Inhibitor
£443.00 - £2740.00
Specifications
Content And Storage | Room temperature |
---|---|
Product Type | mirVana Custom miRNA Inhibitor |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15581124
|
Ambion™
4464086 |
5 nmol |
£588.00
Each |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15501134
|
Ambion™
4464089 |
250 nmol |
£2740.00
250nmol |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15551511
|
Ambion™
4464086 |
5 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically inhibit endogenous miRNAs for the purpose of functional analysis via down-regulation. Use our GeneAssist™ miRNA Workflow Builder to design mimics and inhibitors for any miRNA sequence.Specifications
Room temperature | |
mirVana Custom miRNA Inhibitor |
For Research Use Only. Not for use in diagnostic procedures.