Learn More
Ambion™ Custom mirVana™ miRNA Mimic
Custom mirVana™ miRNA Mimic
£590.00 - £2725.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15531124
|
Ambion™
4464068 |
5 nmol |
£590.00
5nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15541124
|
Ambion™
4464069 |
20 nmol |
£968.00
20nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15551124
|
Ambion™
4464071 |
250 nmol |
£2725.00
250nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. Use our GeneAssist™ miRNA Workflow Builder to design both mimics and inhibitors for any miRNA sequence.For Research Use Only. Not for use in diagnostic procedures.