missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Custom PreAmp Pools
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design.
Brand: Applied Biosystems™ 4478258
174.46 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.
Specifications
Pool of assay primers, -20 C | |
192 array card format | |
Sustainable packaging | |
1 mL |
CUSTOM MIRNA PREAMP POOL (192 A) | |
miRNA | |
miRNA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction