Learn More
Ambion™ Custom RNA
Custom RNA
Brand:  Ambion™ 10620002
17.35 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 10206252
  • £30.43 / 1µmol
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Using our award-winning BLOCK-iT™ RNAi Designer, you can design Custom BLOCK-iT™ siRNA sequences for efficient and specific gene expression knockdowns in any organism. For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNA, 1μmol, Design Custom BLOCK-iT™ siRNA Sequences for Efficient and Specific Gene Expression Knockdowns in any Organism, For Enhanced Target Specificity and Stability in Serum
RNA
1 μmol