Learn More
Ambion™ Custom RNA
Custom RNA
Brand: Ambion™ 100002299
2.72 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Using our award-winning BLOCK-iT™ RNAi Designer, you can design Custom BLOCK-iT™ siRNA sequences for efficient and specific gene expression knockdowns in any organism. For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available.Specifications
Block-iT™ RNAi Express, 100nmol | |
RNA DSL In-vivo | |
100 nmol |