missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom RNA, Stealth, HPLC

Custom RNA, Stealth, HPLC

£40.36 - £400.00

Specifications

Content And Storage Room temperature
Purity HPLC
View More Specs

Products 6
Product Code Brand Description Product Type Price Quantity & Availability  
Product Code Brand Description Product Type Price Quantity & Availability  
10164353
View Documents
Ambion™
100001871
Stealth RNA HPLC, 20nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation Stealth RNA HPLC
£85.00
Each
Please to purchase this item. Need a web account? Register with us today!
10523054
View Documents
Ambion™
100001864
RNA HPLC In-vivo, 20nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation RNA HPLC In-vivo
£227.00
Each
Please to purchase this item. Need a web account? Register with us today!
10328033
View Documents
Ambion™
100001872
Stealth RNA HPLC, 500nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation Stealth RNA HPLC
£102.00
Each
Please to purchase this item. Need a web account? Register with us today!
10667923
View Documents
Ambion™
100001865
RNA HPLC In-vivo, 500nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation RNA HPLC In-vivo
£400.00
Each
Please to purchase this item. Need a web account? Register with us today!
10522504
View Documents
Ambion™
100001870
Stealth RNA HPLC, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation Stealth RNA HPLC
£40.36
Each
Please to purchase this item. Need a web account? Register with us today!
10174283
View Documents
Ambion™
100001863
RNA HPLC In-vivo, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation RNA HPLC In-vivo
£155.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
Specifications

Specifications

Room temperature
HPLC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.