missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Custom Select siRNA
Click to view available options
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
5 nmol
Unit Size:
20nmol
250nmol
40nmol
5nmol
Each
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
Specifications
Specifications
| Content And Storage | Room temperature |
| Description | Custom Select siRNA, 250nmol, HPLC Purity, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities |
| For Use With (Application) | Knocking down gene expression in order to study protein function in a wide range of cell types |
| Product Type | Custom Select siRNA |
| Purity | HPLC |
| Quantity | 250 nmol |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction