missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom Select siRNA, In Vivo Ready

Custom Select siRNA, In Vivo Ready

£1702.00 - £2300.00

Specifications

Content And Storage Room temperature
For Use With (Application) Knocking down gene expression in order to study protein function in a wide range of cell types
Product Type In-vivo Ready Custom Select siRNA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15511064
View Documents Promotion Details
Ambion™
4404006
250 nmol
£1702.00
250nmol
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15581471
View Documents Promotion Details
Ambion™
4404007
1 μmol This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
Specifications

Specifications

Room temperature
Knocking down gene expression in order to study protein function in a wide range of cell types
In-vivo Ready Custom Select siRNA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.