missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Custom Silencer™ siRNA
Custom Silencer™ siRNA
Brand: Ambion™ AM16104
121.94 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.Specifications
Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water | |
siRNA | |
Ambion™, Silencer™ Select | |
Standard | |
Liquid |
Custom siRNA, 20nmol, Liquid, Standard Purity, Water Soluble, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA | |
RNA | |
Custom siRNA | |
20 nmol |
For Research Use Only. Not for use in diagnostic procedures.