Learn More
Applied Biosystems™ Custom TaqMan™ PreAmp Pool
Custom TaqMan™ PreAmp Pool
Brand: Applied Biosystems™ 4441856
225.70 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.Specifications
Custom TaqMan™ PreAmp Pool, 1mL, Custom miRNA Array Designs Can Hold From 12 to 380 Assays, And 1 to 8 Samples On Each Array Design |