Learn More
Invitrogen™ Customer-Defined Anti-miR™ miRNA Inhibitor
Click to view available options
Quantity:
20 nmol
Unit Size:
20nmol
Description
Use our customization tool to design the ideal inhibitor for your miRNA sequence, or select from among predesigned products.
Specifications
Specifications
| Content And Storage | The Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at -20°C. It is guaranteed for 1 year when stored dried (at -20°C.) |
| RNAi Type | miRNA |
| Product Line | Ambion, Anti-miR |
| Product Type | miRNA Inhibitor |
| Quantity | 20 nmol |