Learn More
Fritsch™ Cutting Mill Pulverisette
Brand: Fritsch™ 15.4020.00
Code : 04
Additional Details : Weight : 37.00000kg
Description
All grinding parts can be easily removed for cleaning. Different rotors, various knife geometries and replaceable blades ensure maximum flexibility and durability. For controlling the abrasion properties, the grinding tools are offered in different materials. The optimum airflow ensures fast grinding and secure protection against clogging. The patented Fritsch sample exhauster with cyclone separator (optional) ensures simple feeding, fast throughput and good cooling.Please note that Pulverisette 15 and Pulverisette 19 are intended for sample processing only, not for continuous or heavy duty industrial use.
Ideal for pre- and fine grinding of soft to medium hard as well as fibrous or tough materials. Essential for the areas of plastics, textiles, agriculture and forestry, environmental, construction materials, chemistry, foodstuffs and for sample preparation for RoHS analysis.