Learn More
Agilent BioTek Cuvette Adapter
For use with Microplate Washers and Readers
Brand: Agilent BioTek 7302030
This item is not returnable.
View return policy
Description
- With this adapter up to 4 standard cuvettes can be placed horizontally and read directly in the plate readers - care must be taken that the cuvette is stoppered or sealed
- The adapter has 4 slots for cuvettes, with aperatures designed for several reading position in a standard microplate footprint
Specifications
Cuvette Adapter | |
Agilent BioTek Microplate Washers and Readers | |
Designed for Several Reading Positions in a Standard Microplate Footprint | |
1 Pk. |