missing translation for 'onlineSavingsMsg'
Learn More

CX 546, Tocris Bioscience™

Product Code. 18054292
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

AMPA receptor potentiator

AMPA receptor potentiator. Binds specifically to the agonist bound non-desensitized receptor, most likely destabilizing the desensitized receptor conformation. Enhances cognitive function in rats. Neuroprotective in mice undergoing repeat ketamine anesthesia.

Chemical Identifiers

CAS 215923-54-9
Molecular Formula C14H17NO3
Molecular Weight (g/mol) 247.294
InChI Key LJUNPHMOGNFFOS-UHFFFAOYSA-N
Synonym 1-1,4-benzodioxan-6-ylcarbonyl piperidine, 2,3-dihydrobenzo b 1,4 dioxin-6-yl piperidin-1-yl methanone, unii-pv6yec8983, bdp 17, 2,3-dihydro-1,4-benzodioxin-6-yl piperidin-1-yl methanone, methanone, 2,3-dihydro-1,4-benzodioxin-6-yl-1-piperidinyl, 2,3-dihydro-1,4-benzodioxin-6-yl-1-piperidinylmethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl carbonyl piperidine, 1-2,3-dihydro-1,4-benzodioxine-6-carbonyl piperidine, lopac-c-271
PubChem CID 2890
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-yl(piperidin-1-yl)methanone
SMILES C1CCN(CC1)C(=O)C2=CC3=C(C=C2)OCCO3

Specifications

Quantity 10 mg
Formula Weight 247.29
Percent Purity >99%
Chemical Name or Material CX 546
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'