Learn More
CX3CR1 Monoclonal Antibody (2A9-1), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£97.00 - £246.00
Specifications
Antigen | CX3CR1 |
---|---|
Clone | 2A9-1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15528176
|
Invitrogen
17-6099-41 |
25 Tests |
£97.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15538176
|
Invitrogen
17-6099-42 |
100 Tests |
£246.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This 2A9-1 monoclonal antibody reacts with human CX3CR1, which is the receptor for fractalkine, a transmembrane chemokine of the CX3C family. CX3CR1 is a seven transmembrane G protein-coupled receptor expressed on CD16+ NK cells, T cells (e.g. CD8+, CD4+, and gamma/delta), and monocytes. In non-immune cells, CX3CR1 has been found on osteoclast precursors and microglia. Little to no CX3CR1 surface expression can be detected on B cells and granulocytes. Together, fractalkine and its receptor mediate cell-cell adhesion and chemotaxis of NK cells, T cells, and monocytes. The expression of CX3CR1 has also been correlated with high levels of intracellular perforin and granzyme B. Applications Reported: This 2A9-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2A9-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CX3CR1 Monoclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Flow Cytometry.Specifications
CX3CR1 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Beta chemokine receptor-like 1; CCRL1; chemokine (C-C) receptor-like 1; chemokine (C-X3-C motif) receptor 1; chemokine (C-X3-C) receptor 1; CMKBRL1; CMK-BRL1; CMK-BRL-1; CMKDR1; Cx3c; CX3C chemokine receptor 1; C-X3-C CKR-1; C-X3-C motif chemokine receptor 1; Cx3cr1; Fractalkine receptor; G protein-coupled receptor 13; GPR13; G-protein coupled receptor 13; GPRV28; RBS11; Scyd1; V28 | |
CX3CR1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CX3CR1 |
2A9-1 | |
Flow Cytometry | |
APC | |
Rat | |
Human | |
P49238 | |
1524 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.