missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR4 Mouse anti-Human, Clone: 4B5E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60042-1-IG-20UL
This item is not returnable.
View return policy
Description
CXCR4 may be involved in WHIM syndrome (WHIMS). Immunodeficiency disease characterized by neutropenia, hypogammaglobulinemia and extensive human papillomavirus (HPV) infection. Despite the peripheral neutropenia, bone marrow aspirates from affected individuals contain abundant mature myeloid cells, a condition termed myelokathexis. The disease is caused by mutations affecting the gene represented in this entry.Specifications
CXCR4 | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61073 | |
CXCR4 | |
CXCR4 Fusion Protein Ag1528 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgM |
Western Blot, Flow Cytometry | |
4B5E4 | |
Unconjugated | |
CXCR4 | |
CD184, CXC R4, CXCR 4, CXCR4, CXCR4B, D2S201E, FB22, Fusin, HM89, HSY3RR, LAP3, LCR1, LESTR, SDF 1 receptor, WHIM | |
Mouse | |
Antigen Affinity Chromatography | |
RUO | |
7852 | |
-20°C | |
Liquid |