missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CXorf21 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Brand: Abnova™ H00080231-T01.100uL
This item is not returnable.
View return policy
Specifications
293T | |
CXorf21 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 33.9kDa | |
Transient overexpression cell lysate was tested with anti-CXorf21 antibody (Mfr. No. H00080231-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
NM_025159 | |
pCMV-CXorf21 full-length | |
100 μL | |
Human |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UL CXorf21 293T Cell Transient Overexpression L
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Mercaptoethanol