Learn More
Cytiva Cy™3.5 Amidite
For CyDye™ incorporation at any position in an oligonucleotide during synthesis
Brand:  Cytiva 27-1798-01
Product Code. 10309874
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
CyDye™ fluors combine exceptional photostability with bright and intense signals, resulting in high sensitivity and a highly reproducible performance at a biologically suitable pH range of 3 to 10.
  • Wide selection of CyDye™ fluors for protein labeling for use in ELISA & immunoprecipitation
  • Labeling strategy based on protein characteristics
  • Medium to high-throughput fluorescence imaging
  • Ready-to-use labeled antibodies enable multiplexed assays
  • Streptavidin-conjugated CyDye™ fluors for use in Flow-cytometry and In situ hybridization
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
  • DNA synthesis reagent
Amersham™ CyDye™ fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye™ fluors enables multiplexing without crosstalk. In addition, the CyDye™ flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of Amersham™ CyDye™ fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which together with minimal cross-talk contributes to high accuracy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials
Cy3.5
Cy3.5 Amidite
Solid
Stable
Cy™
100 mg
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1