missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cy3™ Dye-Labeled Pre-miR Negative Control #1
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
Description
Invitrogen™ Cy™3 dye-labeled synthetic miRNA mimic for miRNA functional analysis. 5 nmol is supplied.
Optimized design results in correct strand utilization by RISC
Purified and ready to transfect
Invitrogen™ Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments. The Cy™3 Dye-Labeled Pre-miR™ Negative Control #1 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. It is ideal for monitoring transfection efficiency, as the fluorescent label enables direct observation of the cellular uptake, distribution, and localization of the control. The label is on the 5' end of the molecule, and the oligonucleotide sequence is the same as the Pre-miR™ miRNA Precursor Molecule Negative Control #1 (SKU #AM17110). NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
Invitrogen™ Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments. The Cy™3 Dye-Labeled Pre-miR™ Negative Control #1 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. It is ideal for monitoring transfection efficiency, as the fluorescent label enables direct observation of the cellular uptake, distribution, and localization of the control. The label is on the 5' end of the molecule, and the oligonucleotide sequence is the same as the Pre-miR™ miRNA Precursor Molecule Negative Control #1 (SKU #AM17110). NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
Specifications
Specifications
| Content And Storage | Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C. |
| Format | Tube |
| Control Type | Negative Control |
| RNAi Type | miRNA Mimic |
| For Use With (Application) | miRNA & Non-Coding RNA Analysis |
| Label or Dye | Cy3 |
| Product Line | Ambion, Cy3, Pre-miR |
| Product Type | miRNA Inhibitor |
| Purity | HPLC |
| Quantity | 5 nmol |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction