Learn More
Cytiva Cy5.5 NHS Ester Mono-Reactive CyDye
Use for highly accurate labeling of amine groups. Cytiva Cy5.5 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.
£308.00 - £5893.00
Specifications
Format | Powdered |
---|---|
Product Type | Cy5.5 NHS Ester |
Kit Contents | Foil packs of premeasured dye |
pH Range | 3 to 10 |
Label or Dye | Cy5.5 |
Product Code | Brand | Quantity | Content And Storage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Content And Storage | Price | Quantity & Availability | ||||
10461835
|
Cytiva
PA15604 |
10 mg | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
£1875.00
Each |
Estimated Shipment: 20-11-2024
Log in to see stock. |
||||
10724927
|
Cytiva
PA15602 |
25 mg | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
£5893.00
50mg |
Estimated Shipment: 14-01-2025
Log in to see stock. |
||||
11585085
|
Cytiva
PA15605 |
5 mg | 2°C to 8°C |
£1057.00
5mg |
Estimated Shipment: 14-01-2025
Log in to see stock. |
||||
11575085
|
Cytiva
PA15606 |
25 mg | 2°C to 8°C |
£3414.00
25mg |
Estimated Shipment: 14-01-2025
Log in to see stock. |
||||
10713797
|
Cytiva
PA15601 |
1 mg | 2°C to 8°C |
£308.00
1mg |
In Stock
Log in to see stock. |
||||
Description
CyDye fluors are versatile fluorophores designed for a broad range of applications. CyDye fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye flours enables multiplexing without crosstalk for high accuracy.
Cy5.5 NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.
- Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
- High purity and thorough QC testing generate high levels of chromophore and reactive dye content
- Narrow excitation and emission bands result in discrete signals from each fluor
- Demonstrates larger polarization range than conventional fluors
- Fluorescence lifetime of 2.8 ns
- High fluorescence intensity
- Low non-specific binding
Specifications
Powdered | |
Foil packs of premeasured dye | |
Cy5.5 |
Cy5.5 NHS Ester | |
3 to 10 | |
Blue |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1