missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanide solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£87.50 - £155.00
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | H2O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12992225
|
Fisher Chemical
A 054 100 |
100 mL |
£109.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12902235
|
Fisher Chemical
A 054 250 |
250 mL |
£87.50
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12912235
|
Fisher Chemical
A 054 500 |
500 mL | |||||||
Specifications
1g/L | |
H2O |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-component Standard Solution for Ion chromatography. Cyanides (CN-) 1000mg/l in KOH 0.1% (A 054)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .