missing translation for 'onlineSavingsMsg'
Learn More

Cyanide, standard solution, Specpure™ CN∣- 1000μg/mL

Product Code. 11340448
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11340448

Brand: Thermo Scientific Alfa Aesar 043804.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KCN in 0.1% NaOH

Cyanide, standard solution is used to dissolve metals and their ores. It is also used in clinical chemistry and in waste water treatment facilities to determine the concentration of dissolved and particulate materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula KCN in 0.1% NaOH
MDL Number MFCD00468042

Specifications

Odor Odorless
Density 1g/cm3 at 20°C
Physical Form Liquid
Color Colorless
Solubility Miscible with water
Quantity 100 mL
Concentration or Composition (by Analyte or Components) CN: 1000μg/mL
Chemical Name or Material Cyanide, Standard Solution
Product Identifier
  • Cyanide, standard solution, Specpure, CN 1000µg/ml
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Potassium cyanide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.