Learn More
Cyanogen bromide, Thermo Scientific Chemicals
Description
Cyanogen bromide is used as a reagent in molecular biology, peptide mapping, gold extraction and amino acid sequence analysis. It acts as a coupling reagent for phosphoramidate or pyrophosphate inter-nucleotide bonds in DNA duplexes. It is also used in organic synthesis to prepare cyanamides, dicyanamides, cyanuric bromide, guanidines and hydroxylguanidines. It is involved in the von Braun reaction, it gives disubstituted cyanamides by reacting with tertiary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 50°C to 52°C |
Density | 2.015 |
Boiling Point | 61.4°C |
Quantity | 25 g |
Sensitivity | Light and moisture sensitive |
Solubility Information | Soluble in chloroform,dichloromethane,ethanol,diethyl ether,benzene and acetonitrile. |
Formula Weight | 105.93 |
Physical Form | Crystals |
Chemical Name or Material | Cyanogen bromide |
RUO – Research Use Only