missing translation for 'onlineSavingsMsg'
Learn More
Cyanopropylmethyldichlorosilane, 97%, Thermo Scientific™ Product Code.: 10139649

Cyanopropylmethyldichlorosilane, 97%, Thermo Scientific™

Product Code. 10139649
25 mL, Glass bottle
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10139649

Brand: Thermo Scientific Acros 432250250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1190-16-5
Molecular Formula C5H9Cl2NSi
Molecular Weight (g/mol) 182.12
MDL Number MFCD00019889
InChI Key UIFBMBZYGZSWQE-UHFFFAOYSA-N
Synonym 3-cyanopropyl methyldichlorosilane, 3-cyanopropylmethyldichlorosilane, 4-dichloromethylsilyl-butyronitrile, 3-cyanopropyldichloromethylsilane, butanenitrile, 4-dichloromethylsilyl, unii-a09zd6eimj, 4-dichloromethylsilyl butyronitrile, methylcyanopropyldichlorosilane, 4-dichloro methyl silyl butanenitrile, dichlor-3-kyanpropyl-methylsilan
PubChem CID 14479
IUPAC Name 4-[dichloro(methyl)silyl]butanenitrile
SMILES C[Si](Cl)(Cl)CCCC#N

Specifications

Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 93.0°C to 94.0°C (8.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Quantity 25 mL
Specific Gravity 1.15
Formula Weight 182.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyanopropylmethyldichlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyanopropylmethyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyanopropylmethyldichlorosilane, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.