missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric acid, 98%, Thermo Scientific Chemicals

£14.77 - £118.00

Chemical Identifiers

CAS 108-80-5
Molecular Formula C3H3N3O3
Molecular Weight (g/mol) 129.08
MDL Number MFCD00082990
InChI Key ZFSLODLOARCGLH-UHFFFAOYSA-N
Synonym cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
PubChem CID 7956
ChEBI CHEBI:38028
IUPAC Name 1,3,5-triazinane-2,4,6-trione
SMILES O=C1NC(=O)NC(=O)N1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10184340
View Documents
Thermo Scientific Acros
181020250
25 g Plastic bottle
£14.77
25g
Please to purchase this item. Need a web account? Register with us today!
10635822
View Documents
Thermo Scientific Acros
181022500
250 g Plastic bottle
£35.40
250g
Please to purchase this item. Need a web account? Register with us today!
10701081
View Documents
Thermo Scientific Acros
181020010
1 kg Plastic bottle
£118.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-80-5
129.08
ZFSLODLOARCGLH-UHFFFAOYSA-N
7956
1,3,5-triazinane-2,4,6-trione
C3H3N3O3
MFCD00082990
cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
CHEBI:38028
O=C1NC(=O)NC(=O)N1

Specifications

108-80-5
98%
MFCD00082990
15, 2690
ZFSLODLOARCGLH-UHFFFAOYSA-N
1,3,5-triazinane-2,4,6-trione
7956
129.07
Authentic
Cyanuric acid, 98%
White to Beige
C3H3N3O3
26, 239
cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
O=C1NC(=O)NC(=O)N1
129.08
CHEBI:38028
≥97.5%
Crystalline Powder or Lumps
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

missing translation for 'einecsNumber' : 203-618-0

missing translation for 'rtecsNumber' : XZ1800000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only