missing translation for 'onlineSavingsMsg'
Learn More
Cyanuric acid, 99%, Thermo Scientific Chemicals Product Code.: 11374539

Cyanuric acid, 99%, Thermo Scientific Chemicals

Product Code. 11374539
100 g
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11374539

Brand: Thermo Scientific Alfa Aesar A15447.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cyanuric acid acts as a chlorine stabilizer in swimming pools. It is used as a precursor to N-chlorinated cyanurates, which are used to disinfect water and crosslinking agents for polyurethane resins. It serves as a process regulator. Further, it is used in the diagnostic determination of melamine and related compounds in kidney tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanuric acid acts as a chlorine stabilizer in swimming pools. It is used as a precursor to N-chlorinated cyanurates, which are used to disinfect water and crosslinking agents for polyurethane resins. It serves as a process regulator. Further, it is used in the diagnostic determination of melamine and related compounds in kidney tissue.

Solubility
Soluble in sulfuric acid, dimethylformamide, sodium hydroxide, potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone, benzene, diethyl ether, ethanol and hexane. Insoluble in cold methanol, benzene and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-80-5
Molecular Formula C3H3N3O3
Molecular Weight (g/mol) 129.08
MDL Number MFCD00082990
InChI Key ZFSLODLOARCGLH-UHFFFAOYSA-N
Synonym cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acidShow MoreShow Less
PubChem CID 7956
ChEBI CHEBI:38028
IUPAC Name 1,3,5-triazinane-2,4,6-trione
SMILES O=C1NC(=O)NC(=O)N1

Specifications

Melting Point >360°C
Assay Percent Range 99%
Beilstein 126982
Merck Index 14,2698
Solubility Information Soluble in sulfuric acid,dimethylformamide,sodium hydroxide,potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone,benzene,diethyl ether,ethanol and hexane. Insoluble in cold methanol,benzene and chloroform.
Formula Weight 129.08
Percent Purity 99%
Density 2.5
Quantity 100 g
Chemical Name or Material Cyanuric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242835 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog NumberA15447.0B,A15447.22,A15447.30
Lot NumberFA021907 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15447.0B,A15447.22,A15447.30
Lot Number10191309 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15447.0B,A15447.22,A15447.30
Lot NumberJ19W007 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15447.0B,A15447.22,A15447.30
Lot Number10220508 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15447.0B,A15447.22,A15447.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyanuric acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.