missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CyberScan TB 1000 Turbidity Meters

Product Code. 10724247
Click to view available options
Type:
Lamp Module
Turbidity Meter
Turbidity Meter Calibration Kit
Light Source:
White Light
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10724247

Brand: Thermo Scientific™ 01X265102

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The CyberScan TB1000 series turbidity meters are designed to meet the stringent requirements of today's scientists.

The CyberScan TB1000 series turbidity meters are designed to meet the stringent requirements of today's scientists. TB1000 series meet the performance criteria specified by USEPA method 180.1 for NTU measurement and ISO 7027 (IR version). Ideal for beverage production, drinking water, waste water treatment, petrochemical and electroplating applications.

Features

  • Multi-Point Push-Button Calibration (Up to 3 points) for optimum full-range accuracy of ± 2% full scale
  • Auto-Ranging for fast response and best resolution (up to 0.01 NTU) over wide measurement range
  • Push-Button Calibration is hassle-free, quick and easy
  • Auto-Alert Calibration prompts at user-defined interval to ensure regularly scheduled recalibration for ISO and GLP compliance
  •  RS232 Communication interface to download data to a printer or PC for report documentation  
  • Built-in Real-time Clock stamps stored data and calibration data with time & date - meets standards for GLP purpose
  • Self-Diagnostic with message codes for easy troubleshooting • Pour-Through Assembly Module (optional) allows instantaneous sampling without having to transfer samples into cuvettes thus saving time.
TRUSTED_SUSTAINABILITY

Specifications

Type Turbidity Meter Calibration Kit
Light Source White Light
Automatic Range Selection Yes
For Use With (Equipment) CyberScan TB 1000 - ECTBDIR1000
Description Calibration kit set for CyberScan TB 1000 - ECTBDIR1000 (0.02, 10.0, 1000 NTU)
Resolution 0.01 NTU
Turbidity 0.02, 10.0, 1000 NTU
EPA Approved Methods 180.1
ISO Accepted Methods ISO 7027
compliance-icons
Product Identifier
  • Turbidity standards
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Styrene Divinyl Benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.