Learn More
Contec™ Non-Sterile CyChlor

Provided ready-to-use and is efficacious against bacteria and yeasts in 3 mins. Contec™ Non-Sterile CyChlor is a new broad spectrum disinfectant designed for every day use.
Brand: Contec™ FBT100CC
84.52 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Broad spectrum efficacy in 3 mins saves time spent on biodecontamination.
- Filtered to 0.2 microns and filled in a Grade A environment ensuring the product is free from contamination and particulates
- Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
- No hazard classification so only basic PPE required and no special disposal is necessary
- Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
- Double bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
- Not classed as corrosive and can be used safely in all areas of the cleanroom — this needs a bit of random text deleting from the end

Specifications
Hydrogen Peroxide and Purified Water Blend | |
3 to 6 | |
Non-sterile | |
Not Autoclavable | |
Fluid | |
Limited-use | |
6/Cs. |
Liquid | |
Original | |
Colorless | |
Clear | |
Trigger Spray Bottle | |
No | |
1 L |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
241102939 | Certificate of Quality | 13/02/2025 | FBT100CC |
241002904 | Certificate of Quality | 04/12/2024 | FBT100CC |
240802869 | Certificate of Quality | 24/10/2024 | FBT100CC |
240702815 | Certificate of Quality | 29/08/2024 | FBT100CC |
240602776 | Certificate of Quality | 13/07/2024 | FBT100CC |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Your input is important to us. Please complete this form to provide feedback related to the content on this product.