missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5122143
264.60 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.
Specifications
Cyclin A1 | |
Polyclonal | |
Unconjugated | |
CCNA1 | |
CCNA1; CT146; cyclin A1; cyclin-A1; testicular tissue protein Li 34 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12427, 295052, 8900 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P78396, Q61456, Q6AY13 | |
CCNA1 | |
Ser183-Tyr430 (Accession P78396), with N-terminal His and GST Tag | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cyclin A1 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction