Learn More
Cyclin B2 Mouse anti-Human, CoraLite 647, Clone: 2G8B7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-67726-100UL
Additional Details : Weight : 0.00020kg
Description
The Cyclin B2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cyclin B2. This antibody recognizes human, mouse antigen. The Cyclin B2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™647.
Specifications
Cyclin B2 | |
Monoclonal | |
CoraLite 647 | |
100 μL | |
9133 | |
IgG2a |
Immunofluorescence | |
2G8B7 | |
Mouse | |
Primary | |
Human, Mouse |