Learn More
Cyclin H Mouse anti-Human, Unconjugated, Clone: 2C5G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67065-1-IG-20UL
This item is not returnable.
View return policy
Description
The Cyclin H antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cyclin H. This antibody recognizes human, mouse, rat antigen. The Cyclin H antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Cyclin H | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2C5G11 | |
Unconjugated | |
20 μL | |
902 | |
IgG2a |