missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarbonyl chloride, 98%, Thermo Scientific Chemicals

 

£84.40 - £280.00

Chemical Identifiers

CAS 5006-22-4
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001319
InChI Key JFWMYCVMQSLLOO-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
PubChem CID 78705
IUPAC Name cyclobutanecarbonyl chloride
SMILES C1CC(C1)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419836
View Documents
Thermo Scientific Alfa Aesar
B20317.09
10 g
£84.40
10g
Please to purchase this item. Need a web account? Register with us today!
11429836
View Documents
Thermo Scientific Alfa Aesar
B20317.18
50 g
£280.00
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

5006-22-4
118.56
JFWMYCVMQSLLOO-UHFFFAOYSA-N
78705
C1CC(C1)C(=O)Cl
C5H7ClO
MFCD00001319
cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
cyclobutanecarbonyl chloride

Specifications

5006-22-4
145°C
Stench
1.455
UN2920
Moisture Sensitive
JFWMYCVMQSLLOO-UHFFFAOYSA-N
cyclobutanecarbonyl chloride
78705
98%
1.039
37°C (99°F)
C5H7ClO
MFCD00001319
956624
cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
C1CC(C1)C(=O)Cl
118.56
118.56
Cyclobutanecarbonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 225-680-8

missing translation for 'tsca' : No

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only