missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanone, 98+%, Thermo Scientific Chemicals

Product Code. 10560191
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-95-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00001332
InChI Key SHQSVMDWKBRBGB-UHFFFAOYSA-N
Synonym unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
PubChem CID 14496
IUPAC Name cyclobutanone
SMILES O=C1CCC1

Specifications

Melting Point -50.9°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 99.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C4H6(=O)
Refractive Index 1.4205 to 1.4225
Quantity 1 g
Beilstein 07,5
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 70.09
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Cyclobutanone
Show More Show Less
compliance-icons
Product Identifier
  • Cyclobutanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'