missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutyl bromide, 97%, pure, Thermo Scientific Chemicals

Product Code. 10521161
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4399-47-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.01
MDL Number MFCD00001317
InChI Key KXVUSQIDCZRUKF-UHFFFAOYSA-N
Synonym cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
PubChem CID 78110
IUPAC Name bromocyclobutane
SMILES C1CC(C1)Br

Specifications

Color Colorless
Density 1.4300g/mL
Boiling Point 108.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C4H7Br
Refractive Index 1.4776 to 1.4797
Quantity 1 g
Specific Gravity 1.43
Solubility Information Solubility in water: immiscible
Formula Weight 135.01
Percent Purity 97%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cyclobutyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'