Learn More
Cyclobutylamine hydrochloride, 97%, Thermo Scientific Chemicals
£63.90 - £217.08
Chemical Identifiers
CAS | 6291-01-6 |
---|---|
Molecular Formula | C4H10ClN |
Molecular Weight (g/mol) | 107.58 |
MDL Number | MFCD00034953 |
InChI Key | NFAZOGXQOWEWBM-UHFFFAOYSA-N |
Synonym | cyclobutanamine hydrochloride, cyclobutylamine hydrochloride, aminocyclobutane hcl, cyclobutanamine, hydrochloride, cyclobutanaminium chloride, cyclobutylamine hcl, acmc-209wdt, aminocyclobutane hydrochloride, cyclobutylamine hydrochloric acid, cyclobutylamine hydrochloride salt |
PubChem CID | 12618374 |
IUPAC Name | cyclobutanamine;hydrochloride |
SMILES | C1CC(C1)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11461020
|
Thermo Scientific Alfa Aesar
B25400.06 |
5 g |
£63.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11471020
|
Thermo Scientific Alfa Aesar
B25400.14 |
25 g |
£217.08
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool, dry conditions, in well sealed container. Incompatible with oxidizing agents.
Chemical Identifiers
6291-01-6 | |
107.58 | |
NFAZOGXQOWEWBM-UHFFFAOYSA-N | |
12618374 | |
C1CC(C1)N.Cl |
C4H10ClN | |
MFCD00034953 | |
cyclobutanamine hydrochloride, cyclobutylamine hydrochloride, aminocyclobutane hcl, cyclobutanamine, hydrochloride, cyclobutanaminium chloride, cyclobutylamine hcl, acmc-209wdt, aminocyclobutane hydrochloride, cyclobutylamine hydrochloric acid, cyclobutylamine hydrochloride salt | |
cyclobutanamine;hydrochloride |
Specifications
6291-01-6 | |
C4H10ClN | |
Hygroscopic | |
Soluble in water. | |
C1CC(C1)N.Cl | |
107.58 | |
107.58 | |
Cyclobutylamine hydrochloride |
180°C to 183°C | |
MFCD00034953 | |
cyclobutanamine hydrochloride, cyclobutylamine hydrochloride, aminocyclobutane hcl, cyclobutanamine, hydrochloride, cyclobutanaminium chloride, cyclobutylamine hcl, acmc-209wdt, aminocyclobutane hydrochloride, cyclobutylamine hydrochloric acid, cyclobutylamine hydrochloride salt | |
NFAZOGXQOWEWBM-UHFFFAOYSA-N | |
cyclobutanamine;hydrochloride | |
12618374 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 228-540-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only