missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Cyclodextrin hydrate, 98+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Melting Point | 278°C |
Color | White |
Loss on Drying | 11% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.5% max. Linear oligomers (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 14, 2718 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'