Learn More
Cycloheptane, 99%, Thermo Scientific Chemicals
£26.10 - £281.00
Chemical Identifiers
CAS | 291-64-5 |
---|---|
Molecular Formula | C7H14 |
Molecular Weight (g/mol) | 98.19 |
MDL Number | MFCD00004147 |
InChI Key | DMEGYFMYUHOHGS-UHFFFAOYSA-N |
Synonym | heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid |
PubChem CID | 9265 |
IUPAC Name | cycloheptane |
SMILES | C1CCCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10457100
|
Thermo Scientific Acros
111010050 |
5 mL | Glass bottle |
£26.10
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10259683
|
Thermo Scientific Acros
111010250 |
25 mL | Glass bottle |
£86.10
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10030830
|
Thermo Scientific Acros
111011000 |
100 mL | Glass bottle |
£281.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
291-64-5 | |
98.19 | |
DMEGYFMYUHOHGS-UHFFFAOYSA-N | |
9265 | |
C1CCCCCC1 |
C7H14 | |
MFCD00004147 | |
heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid | |
cycloheptane |
Specifications
291-64-5 | |
Colorless | |
118.4°C | |
Authentic | |
C7H14 | |
1.4440 to 1.446 | |
05,29 | |
heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid | |
DMEGYFMYUHOHGS-UHFFFAOYSA-N | |
cycloheptane | |
9265 | |
99% | |
Cycloheptane |
-12°C | |
0.8100g/mL | |
6°C | |
98.5% min. (GC) | |
C7H14 | |
MFCD00004147 | |
0.81 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol | |
C1CCCCCC1 | |
98.19 | |
98.19 | |
Liquid |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-030-2
- Cycloheptane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only