missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanecarboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 10654365
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1460-16-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00004152
InChI Key VZFUCHSFHOYXIS-UHFFFAOYSA-N
Synonym cycloheptane carboxylic acid, cycloheptanoic acid, cycloheptanecarboxylicacid, cyclohepatan carboxylic acid, acmc-1c1rc, sampl4, o9, vzfuchsfhoyxis-uhfffaoysa, cycloheptanecarboxylic acid
PubChem CID 15091
ChEBI CHEBI:29564
IUPAC Name cycloheptanecarboxylic acid
SMILES OC(=O)C1CCCCCC1

Specifications

Color Colorless
Density 1.0350g/mL
Boiling Point 135.0°C to 138.0°C (9.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C7H13CO2H
Refractive Index 1.4710 to 1.4730 (20°C, 589nm)
Quantity 5 g
Beilstein 09, 12
Specific Gravity 1.035
Formula Weight 142.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cycloheptanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • Cycloheptanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'