Learn More
Cycloheptanone, 98+%, Thermo Scientific Chemicals
£31.70 - £142.00
Chemical Identifiers
CAS | 502-42-1 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.172 |
MDL Number | MFCD00004159 |
InChI Key | CGZZMOTZOONQIA-UHFFFAOYSA-N |
Synonym | suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane |
PubChem CID | 10400 |
IUPAC Name | cycloheptanone |
SMILES | C1CCCC(=O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11485453
|
Thermo Scientific Alfa Aesar
A13421.18 |
50 g |
£31.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495453
|
Thermo Scientific Alfa Aesar
A13421.30 |
250 g |
£142.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cycloheptanone is used as a precursor for the synthesis of pharmaceuticals. A spasmolytic agent and vasodilator is produced from this chemical. They are also used in certain stereospecific enzymatic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCycloheptanone is used as a precursor for the synthesis of pharmaceuticals. A spasmolytic agent and vasodilator is produced from this chemical. They are also used in certain stereospecific enzymatic reactions.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents,strong bases, reducing agents.
Chemical Identifiers
502-42-1 | |
112.172 | |
CGZZMOTZOONQIA-UHFFFAOYSA-N | |
10400 | |
C1CCCC(=O)CC1 |
C7H12O | |
MFCD00004159 | |
suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane | |
cycloheptanone |
Specifications
502-42-1 | |
0.956 | |
55°C (131°F) | |
C7H12O | |
MFCD00004159 | |
969823 | |
suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane | |
CGZZMOTZOONQIA-UHFFFAOYSA-N | |
cycloheptanone | |
10400 | |
99% |
-21°C | |
179°C to 181°C | |
Sweet | |
1.461 | |
UN1224 | |
14,2722 | |
It is insoluble in water. | |
C1CCCC(=O)CC1 | |
112.172 | |
112.17 | |
Cycloheptanone |
Safety and Handling
GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P374-P380-P501c
H226-H302+H332-H318
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 207-937-6
missing translation for 'rtecsNumber' : GU3325000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only