missing translation for 'onlineSavingsMsg'
Learn More

(Cycloheptatriene)molybdenum tricarbonyl, 99%, Thermo Scientific™

Product Code. 11343638
0.5 g
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-77-8
Molecular Formula C10H11MoO3
Molecular Weight (g/mol) 275.15
MDL Number MFCD00004160
InChI Key ZWJSTNVCXCHPHF-UHFFFAOYNA-N
Synonym cycloheptatriene molybdenum tricarbonyl, cycloheptatrienemolybdenum tricarbonyl, tris carbon monoxide cycloheptatriene molybdenum

Specifications

Melting Point 100°C to 101°C (decomposition)
Quantity 0.5 g
UN Number UN3466
Sensitivity Air Sensitive
Solubility Information It is insoluble in water.
Formula Weight 272.11
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material (Cycloheptatriene)molybdenum tricarbonyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Cycloheptatriene)molybdenum tricarbonyl, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.