missing translation for 'onlineSavingsMsg'
Learn More

(Cycloheptatriene)molybdenum tricarbonyl, 99%, Thermo Scientific™

Product Code. 11353638
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-77-8
Molecular Formula C10H11MoO3
Molecular Weight (g/mol) 275.15
MDL Number MFCD00004160
InChI Key ZWJSTNVCXCHPHF-UHFFFAOYNA-N
Synonym cycloheptatriene molybdenum tricarbonyl, cycloheptatrienemolybdenum tricarbonyl, tris carbon monoxide cycloheptatriene molybdenum

Specifications

Melting Point 100°C to 101°C (decomposition)
Quantity 2 g
UN Number UN3466
Sensitivity Air Sensitive
Solubility Information It is insoluble in water.
Formula Weight 272.11
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material (Cycloheptatriene)molybdenum tricarbonyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'