Learn More
(Cycloheptatriene)molybdenum tricarbonyl, 99%, Thermo Scientific™
£85.69 - £243.00
Chemical Identifiers
CAS | 12125-77-8 |
---|---|
Molecular Formula | C10H11MoO3 |
Molecular Weight (g/mol) | 275.15 |
MDL Number | MFCD00004160 |
InChI Key | ZWJSTNVCXCHPHF-UHFFFAOYNA-N |
Synonym | cycloheptatriene molybdenum tricarbonyl, cycloheptatrienemolybdenum tricarbonyl, tris carbon monoxide cycloheptatriene molybdenum |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
12125-77-8 | |
275.15 | |
ZWJSTNVCXCHPHF-UHFFFAOYNA-N |
C10H11MoO3 | |
MFCD00004160 | |
cycloheptatriene molybdenum tricarbonyl, cycloheptatrienemolybdenum tricarbonyl, tris carbon monoxide cycloheptatriene molybdenum |
Specifications
12125-77-8 | |
C10H11MoO3 | |
UN3466 | |
cycloheptatriene molybdenum tricarbonyl, cycloheptatrienemolybdenum tricarbonyl, tris carbon monoxide cycloheptatriene molybdenum | |
ZWJSTNVCXCHPHF-UHFFFAOYNA-N | |
272.11 | |
Crystalline |
100°C to 101°C (decomposition) | |
MFCD00004160 | |
Air Sensitive | |
It is insoluble in water. | |
275.15 | |
99% | |
(Cycloheptatriene)molybdenum tricarbonyl |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 235-197-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures